missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SULT1A2 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93418-0.1ml
This item is not returnable.
View return policy
Description
SULT1A2 Polyclonal antibody specifically detects SULT1A2 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| SULT1A2 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| Aryl sulfotransferase 2, arylamine sulfotransferase, EC 2.8.2, EC 2.8.2.1, HAST4, MGC142287, MGC142289, Phenol sulfotransferase 2, phenolic-metabolizing (P) form of PST, phenol-preferring phenol sulfotransferase2, Phenol-sulfating phenol sulfotransferase 2, P-PST 2, ST1A2, STP2P-PST, sulfotransferase 1A2, sulfotransferase family, cytosolic, 1A, phenol-preferring, member 2, thermostable phenol sulfotransferase, TSPST2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 201-295 of human SULT1A2 (NP_001045.1). KREIQKILEFVGRSLPEETVDLMVEHTSFKEMKKNPMTNYTTVRREFMDHSISPFMRKGMAGDWKTTFTVAQNERFDADYAEKMAGCSLSFRSEL | |
| 0.1 mL | |
| Cell Biology, Endocrinology, Signal Transduction | |
| 6799 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction