missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SULT1A1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10911-2-AP-150UL
This item is not returnable.
View return policy
Description
Sulfotransferase enzymes catalyze the sulfate conjugation of many hormones, neurotransmitters, drugs, and xenobiotic compounds. These cytosolic enzymes are different in their tissue distributions and substrate specificities. The gene structure (number and length of exons) is similar among family members. This gene encodes one of two phenol sulfotransferases with thermostable enzyme activity. Multiple alternatively spliced variants that encode two isoforms have been identified for this gene.Specifications
| SULT1A1 | |
| Polyclonal | |
| Unconjugated | |
| SULT1A1 | |
| Aryl sulfotransferase 1, HAST1/HAST2, P PST, P PST 1, Phenol sulfotransferase 1, PST, ST1A1, ST1A3, STP, STP1, Sulfotransferase 1A1, SULT1A1, Ts PST, TSPST1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 20887, 6817, 83783 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
| 0.2 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P17988, P50225, P52840 | |
| SULT1A1 | |
| SULT1A1 Fusion Protein Ag1344 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction