missing translation for 'onlineSavingsMsg'
Learn More

Sulphuric Acid Solution, EU Pharmacopoeia Grade, Fisher Chemical™

Product Code. 12980894
Click to view available options
Quantity:
1000 mL
Packaging:
1000mL
Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Pharmacopoeia reagents are intended for use in pharmacopoeia testing and monographs, produced under a GMP compliant ISO 9001 quality management system according to the relevant pharmacopoeia specification, using the pharmacopoeia reagent quality components and water meeting the relevant pharmacopoeia monograph requirement for Purified Water. Some pharmacopoeia standards are also accredited according to ISO 17025 and ISO Guide 43.

Specifications

Packaging 1000mL
Quantity 1000 mL
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'