missing translation for 'onlineSavingsMsg'
Learn More

Sulphuric Acid 2.5 M (5.0 N) Analytical Volumetric Solution (AVL), Reagecon
BIOPHARMA

Product Code. 11523012 Shop All Reagecon Products
Click to view available options
Quantity:
1 L
5 L
Packaging:
HDPE Bottle GL45
HDPE Jerrycan
Unit Size:
1 L
5 L
This item is not returnable. View return policy

Product Code. 11523012

Brand: Reagecon SU22501

Please to purchase this item. Need a web account? Register with us today!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Fully factorized, high purity, stable, certified product, developed and tested for acid/base titrations.

  • Titrant, Standard Titrant, Standard Solution, Standardized Solution, Reference Standard, Reference Material
  • Accurate, stable, reproducible and fully factorized.
  • Titrations performed with this product, exhibit defined stoichiometry, are quantitative, establish an equilibrium that is definitive and fast and provide unambiguous results.
  • Stability is proven (either opened or unopened) for the shelf life of this product.
  • Certified by Reagecon’s ISO 17025 Accredited Test Method (INAB Ref: 264T)
  • Certified to a specification of ± 0.1%.
  • Product is a reagent of known concentration, that is dispensed from a burette or other dispensing apparatus to a sample (analyte), until a reaction between the two liquids is judged to be complete.
  • May be used for manual titrations, or automatic titrations, to determine the end points by detection of any of several properties.
  • Acid/base titrations, also called neutralization titrations, are used to determine either the amount of acid/base in a sample or the substance that can be converted to an acid/base.

Spezifikation

Concentration 2.5 M (5.0 N)
Packaging HDPE Bottle GL45
Quantity 1 L
Physical Form Liquid
compliance-icons
Produktidentifikator
  • sulphuric acid 5.0N (2.5M)
Signalwort
  • Danger
Gefahrenkategorie
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 A
Gefahrenhinweis
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Sicherheitshinweis
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Ergänzende Informationen
  • MIXTURE LIST-Contains : sulphuric acid
Berichtigung von Produktinhalten

Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.

Name des Produkts

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.

Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt