Learn More
Sulphuric Acid 1.0 M (2.0 N) Analytical Volumetric Solution (AVL), Reagecon
Fully factorized, high purity, stable, certified product, developed and tested for acid/base titrations.
Brand: Reagecon SU21005
Code : NEW
Additional Details : CAS Number : 7664-93-9 Weight : 5.42000kg Transport : UN number : 2796 Chem class : 8 Pack group : II
Description
- Titrant, Standard Titrant, Standard Solution, Standardized Solution, Reference Standard, Reference Material
- Accurate, stable, reproducible and fully factorized.
- Titrations performed with this product, exhibit defined stoichiometry, are quantitative, establish an equilibrium that is definitive and fast and provide unambiguous results.
- Stability is proven (either opened or unopened) for the shelf life of this product.
- Certified by Reagecon’s ISO 17025 Accredited Test Method (INAB Ref: 264T)
- Certified to a specification of ± 0.1%.
- Product is a reagent of known concentration, that is dispensed from a burette or other dispensing apparatus to a sample (analyte), until a reaction between the two liquids is judged to be complete.
- May be used for manual titrations, or automatic titrations, to determine the end points by detection of any of several properties.
- Acid/base titrations, also called neutralization titrations, are used to determine either the amount of acid/base in a sample or the substance that can be converted to an acid/base.
Specifications
1.0 M (2.0 N) | |
HDPE Jerrycan | |
5 L | |
Liquid |
Safety and Handling
- SULPHURIC ACID 2.0N (1.0M)
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
Precautionary Statement
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P362+P364-Take off contaminated clothing and wash it before reuse.
Supplemental information
- MIXTURE LIST-Contains : sulphuric acid