missing translation for 'onlineSavingsMsg'
Learn More

Sulphuric Acid 0.05 M according to European Pharmacopoeia (EP), Reagecon
BIOPHARMA

Product Code. 12862503 Shop All Reagecon Products
Change view
Click to view available options
Quantity:
1 L
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12862503 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12862503 Supplier Reagecon Supplier No. 3008000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Fully factorized, high-purity, stable product developed and tested for titrations.

  • European Pharmacopoeia (EP) Reference Standard, European Pharmacopoeia (EP) Reference Material
  • Accurate, stable, reproducible and fully factorized.
  • Titrations performed with this product, exhibit defined stoichiometry, are quantitative, establish an equilibrium that is definitive and fast and provide unambiguous results.
  • Stability is proven (either opened or unopened) for the shelf life of this product.
  • Certified to a specification of ± 0.2%.
  • Certified by Reagecon’s ISO 17025 Accredited Test Method (INAB Ref: 264T)
  • Product is a reagent of known concentration, that is dispensed from a burette or other dispensing apparatus to a sample (analyte), until a reaction between the two liquids is judged to be complete.
  • May be used for manual titrations, or automatic titrations, to determine the end points by detection as detailed in several EP monographs.
  • NIST traceable.

Specifications

Description Accuracy ± 0.2%
Packaging HDPE Bottle GL45
Physical Form Liquid
Concentration 0.05 M
Technique Titration, EP Monographs
Quantity 1 L
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.