missing translation for 'onlineSavingsMsg'
Learn More

Sulphur Standard for ICP, ICP-MS 1000 μg/mL (1000 ppm) in Water (H2O), Reagecon
BIOPHARMA

Product Code. 11489029 Shop All Reagecon Products
Click to view available options
Quantity:
250 mL
Unit Size:
250mL
This item is not returnable. View return policy

Product Code. 11489029

Brand: Reagecon PS2B7

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Manufactured from ultra-pure metal (99.999%) tested by an ISO17025 Accredited Test Method (INAB Ref: 264T) and raw materials assayed by titration and ICP-MS prior to manufacture.

  • ICP, ICP-MS Reference Standard, ICP, ICP-MS Reference Materia
  • ISO/IEC 17025 for the testing of ICP/ICP-MS Standards using ICP-MS (INAB Ref: 264T)
  • ISO/IEC 17025 for calibration of laboratory balances (INAB Ref: 265C)
  • ISO/IEC 17025 for temperature calibration in the range of -196 - +1200°C (INAB Ref: 265C)
  • ISO/IEC 17025 for density measurement (INAB Ref: 264T) using Oscillating U-Tube and Bingham Pycnometry
  • ISO/IEC 17025 for certain complexometric titrations (INAB Ref: 264T)
  • GHS/OSHA compliant, REACH etc.
  • This product is produced in a highly controlled cleanroom (ISO 7) environment, using ultra-pure water (specially treated for the production of mass spectroscopy standards), triple distilled acids and triple leached pre-cleaned bottles.
  • The titrimetry and spectroscopy are controlled and calibrated using separate Certified Reference Materials (CRM’s), so the product comes with two layers of traceability including NIST Standards where available.
  • All of the uncertainties of measurement for this product are calculated according to Eurachem/CITAC guidelines and reported as expanded uncertainties at the 95% confidence level.

Specifications

Description Matrix: H2O
Packaging HDPE Triple-leached
Analyte or Component Names Sulphur
Physical Form Liquid
Concentration 1000 µg/mL (1000 ppm)
Technique ICP, ICP-MS, AA, IC, XRF and other cation measurement techniques
Element Sulphur
Quantity 250 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.