missing translation for 'onlineSavingsMsg'
Learn More

Sulfur powder, sublimed, -100 mesh, 99.5%, Thermo Scientific Chemicals

Catalog Number 11357728
Click to view available options
Quantity:
500 g
2 kg
This item is not returnable. View return policy
7704-34-9
S
32.06
MFCD00085316
NINIDFKCEFEMDL-UHFFFAOYSA-N
polymeric, lac, ad usum externum, acmc-20a47y, rubbervulcanizationagentis ot
5362487
CHEBI:26833
sulfur
[S]
This item is not returnable. View return policy
7704-34-9
S
32.06
MFCD00085316
NINIDFKCEFEMDL-UHFFFAOYSA-N
polymeric, lac, ad usum externum, acmc-20a47y, rubbervulcanizationagentis ot
5362487
CHEBI:26833
sulfur
[S]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Yellow
Physical Form Sublimed Powder
Quantity 2 kg
Assay Percent Range 99.50%
Ignition Point 248°C (478°F)
Odor Weak
Appearance Pale yellow powder
Chemical Name or Material Sulphur

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'