Learn More
Palintest™ Sulphate Test Kit
Specifications
Specifications
| No. of Tests | 50 |
| Quantity | 50 Pack |
| Range | 0 to 200mg/L |
| Sufficient For | 50 Tests |
| Type | Sulphate Test Kit |
Safety and Handling
Product Identifier
- sulphate (turbidity) photometry tablets
- Warning
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- H302-Harmful if swallowed.
- H319-Causes serious eye irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P330-Rinse mouth.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- P264-Wash contaminated skin thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- MIXTURE LIST-Contain: adipic acid, bariumchloride, ethylenediaminetetraacetic acid di sodium