missing translation for 'onlineSavingsMsg'
Learn More

Sulphanilamide, TRC

Product Code. 17973915
Change view
Click to view available options
Quantity:
50 mg
1 g
10 g
Unit Size:
10g
1g
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17973915 10 g 10g
17942765 50 mg 50mg
17911565 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17973915 Supplier Toronto Research Chemicals Supplier No. TRCS68910010G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63-74-1
Molecular Formula C6 H8 N2 O2 S
Molecular Weight (g/mol) 172.2
Synonym Sulfanilamide, 4-Aminobenzenesulphonamide, Sulphanilamide, Sulfadimidine Imp. D (Pharmeuropa), Sulfacetamide Sodium Imp. A (EP), Sulfaguanidine Imp. A (EP), Sulfadiazine Imp. D (Pharmeuropa), Sulfamethoxazole Imp. E (EP), Benzenesulfonamide, 4-amino- (9CI, ACI), 4-Aminobenzenesulfonamide (ACI), Sulfanilamide (8CI), (4-(Aminosulfonyl)phenyl)amine, 1162F, 4-(Aminosulfonyl)aniline, 4-Aminobenzene-1-sulfonamide, 4-Aminobenzenesulfonic acid amide, 4-Aminophenylsulfonamide, 4-Sulfamoylaniline, A-349, Albexan, Albosal, Ambeside, Antistrept, Astreptine, Astrocid, Azol, Bacteramid, Bactesid, Collomide, Colsulanyde, Copticide, Deseptyl, Dipron, Ergaseptine, Erysipan, Estreptocida, Exoseptoplix, F 1162, Fourneau 1162, Gerison, Gombardol, Infepan, Lusil, Lysococcine, Neococcyl, NSC 7618, Orgaseptine, p-Aminobenzenesulfonamide, p-Aminophenylsulfonamide, p-Anilinesulfonamide, p-Sulfamidoaniline, p-Sulfamoylaniline, PABS, Prontalbin, Prontosil Album, Prontosil I, Prontosil white, Prontylin, Pronzin Album, Proseptal, Proseptine, Proseptol, Pysococcine, Ro 1-3354, Rubiazol A, Sanamid, Septamide Album, Septanilam, Septinal, Septolix, Septoplex, Septoplix, Stopton Album, Stramid, Strepamide, Strepsan, Streptagol, Streptamid, Streptasol, Streptocid, Streptocid album, Streptocide, Streptocide (white), Streptoclase, Streptocom, Strepton, Streptopan, Streptosil, Streptozol, Streptozone, Sulfamidyl, Sulfamine, Sulfana, Sulfanalone, Sulfanidyl, Sulfanil, Sulfanilamide Vaginal Cream, Sulfocidine, Sulfonamid, Sulfonamide, Sulfonamide P, Sulfonylamide, Sulphanilamide, Sulphonamide, Therapol, Tolder, White streptocide
IUPAC Name 4-aminobenzenesulfonamide
SMILES Nc1ccc(cc1)S(=O)(=O)N

Specifications

Quantity 10 g
InChI Formula InChI=1S/C6H8N2O2S/c7-5-1-3-6(4-2-5)11(8,9)10/h1-4H,7H2,(H2,8,9,10)
Formula Weight 172.0306
Percent Purity >95%
Chemical Name or Material Sulfanilamide
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.