missing translation for 'onlineSavingsMsg'
Learn More

Sulindac Sulfide Methyl Ester, TRC

Product Code. 30412393
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30412393 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30412393 Supplier Toronto Research Chemicals Supplier No. TRCS69931525MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sulindac Sulfide Methyl Ester
CAS 106647-49-8
Molecular Formula C21H19FO2S
Quantity 25 mg
Synonym (1Z)-5-Fluoro-2-methyl-1-[[4-(methylthio)phenyl]methylene]-1H-indene-3-acetic Acid Methyl Ester, (Z)-5-Fluoro-2-methyl-1-[[4-(methylthio)phenyl]methylene]-1H-indene-3-acetic Acid Methyl Ester
InChI Formula InChI=1S/C21H19FO2S/c1-13-18(10-14-4-7-16(25-3)8-5-14)17-9-6-15(22)11-20(17)19(13)12-21(23)24-2/h4-11H,12H2,1-3H3/b18-10-
SMILES COC(=O)CC1=C(C)\C(=C\c2ccc(SC)cc2)\c3ccc(F)cc13
IUPAC Name methyl 2-[(3Z)-6-fluoro-2-methyl-3-[(4-methylsulfanylphenyl)methylidene]inden-1-yl]acetate
Molecular Weight (g/mol) 354.44
Formula Weight 354.109
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.