missing translation for 'onlineSavingsMsg'
Learn More

Sulfuryl chloride, 98.5%, Thermo Scientific Chemicals

Catalog Number 10113900
32.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.


Click to view available options
Quantity:
25 g
1 kg
2.5 kg
Packaging:
Glass bottle
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
7791-25-5
Cl2O2S
134.96
MFCD00011451
YBBRCQOCSYXUOC-UHFFFAOYSA-N
sulfuryl chloride, sulfonyl chloride, sulphuryl dichloride, sulphuryl chloride, sulfonyl dichloride, sulfuric dichloride, sulfuric oxychloride, sulfurylchlorid, caswell no. 816, sulfurylchloride
24648
CHEBI:29291
sulfuryl dichloride
ClS(Cl)(=O)=O
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
7791-25-5
Cl2O2S
134.96
MFCD00011451
YBBRCQOCSYXUOC-UHFFFAOYSA-N
sulfuryl chloride, sulfonyl chloride, sulphuryl dichloride, sulphuryl chloride, sulfonyl dichloride, sulfuric dichloride, sulfuric oxychloride, sulfurylchlorid, caswell no. 816, sulfurylchloride
24648
CHEBI:29291
sulfuryl dichloride
ClS(Cl)(=O)=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent
TRUSTED_SUSTAINABILITY
Boiling Point 69.1°C
Melting Point -54.1°C
Color Colorless to Yellow
Physical Form Liquid
Quantity 1 kg
Assay Percent Range 98% min. (ex Cl) (Argentometry)
Linear Formula SO2Cl2
Fieser 01,1128; 02,394; 03,276; 04,474; 05,641; 06,561; 10,375; 13,284; 14,291; 16,311
Merck Index 15,911
Solubility Information Solubility in water: reacts. Other solubilities: soluble in glacial acetic acid
Formula Weight 134.96
Percent Purity 98.5%
Packaging Glass bottle
Refractive Index 1.443
Specific Gravity 1.667
Density 1.6670g/mL
Chemical Name or Material Sulfuryl chloride, 98.5%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Sulfuryl chloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H335-May cause respiratory irritation.
  • H314-Causes severe skin burns and eye damage.
  • H330-Fatal if inhaled.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'