missing translation for 'onlineSavingsMsg'
Learn More

Sulfuryl chloride, 1M soln. in dichloromethane

Product Code. 15414557
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15414557 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 15414557 Supplier Thermo Scientific Alfa Aesar Supplier No. H31899.AP
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 7791-25-5 | Cl2O2S | 134.96 g/mol

Sulfuryl chloride is often used as a source of chlorine. is widely used as a reagent in the conversion of C-H ? C-Cl adjacent to activating substituent such as carbonyls and sulfoxides. It also chlorinates alkanes, alkenes, alkynes, aromatics, ethers (such as tetrahydrofuran) and epoxides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sulfuryl chloride is often used as a source of chlorine. is widely used as a reagent in the conversion of C-H → C-Cl adjacent to activating substituent such as carbonyls and sulfoxides. It also chlorinates alkanes, alkenes, alkynes,
TRUSTED_SUSTAINABILITY

Specifications

CAS 7791-25-5
Quantity 500 mL
Molecular Formula Cl2O2S
Linear Formula SO2Cl2
MDL Number MFCD00011451
UN Number UN2922
Synonym sulfuryl chloride, sulfonyl chloride, sulphuryl dichloride, sulphuryl chloride, sulfonyl dichloride, sulfuric dichloride, sulfuric oxychloride, sulfurylchlorid, caswell no. 816, sulfurylchloride
Solubility Information Reacts with water.
InChI Key YBBRCQOCSYXUOC-UHFFFAOYSA-N
SMILES ClS(Cl)(=O)=O
IUPAC Name sulfuryl dichloride
Molecular Weight (g/mol) 134.96
PubChem CID 24648
ChEBI CHEBI:29291
Formula Weight 134.96
Concentration or Composition (by Analyte or Components) 1M soln. in dichloromethane
Sensitivity Moisture sensitive
Density 1.352
Chemical Name or Material Sulfuryl chloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.