missing translation for 'onlineSavingsMsg'
Learn More

Sulfuric acid solution, For the determination of fluorine, for textile and cream analysis, 72%, Honeywell™

Product Code. 15680520 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
Packaging:
Poly Bottle
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15680520 1 L Poly Bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Product Code. 15680520 Supplier Honeywell Research Chemicals Supplier No. 006471L

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

for the determination of fluorine, for textile and cream analysis, 72%

  • We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
    Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
    Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
TRUSTED_SUSTAINABILITY

Specifications

CAS 7664-93-9
Boiling Point 110°C
Density 1.290 g/cm3
Packaging Poly Bottle
Quantity 1 L
Molecular Formula H2O4S
Linear Formula H2SO4
MDL Number MFCD00064589
UN Number UN1830
Beilstein 2037554
Synonym oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
SMILES OS(O)(=O)=O
IUPAC Name sulfuric acid
Molecular Weight (g/mol) 98.07
PubChem CID 1118
ChEBI CHEBI:26836
Formula Weight 98.08
Percent Purity ≥72%
Chemical Name or Material Sulfuric acid solution, For the determination of fluorine, for textile and cream analysis, 72%
Show More Show Less
compliance-icons
Product Identifier
  • Sulfuric acid solution
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: sulphuric acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.