missing translation for 'onlineSavingsMsg'
Learn More

Sulfuric acid solution, 0.25M, Reag. Ph. Eur., Solstice

Product Code. 15664990 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
1 L
5 L
10 L
4 x 5 L
6 x 1 L
Unit Size:
1 L
10 L
5 L
Pack of 4
Pack of 6
Packaging:
Plastic Bottle
VOLPAC™ CONTAINER
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize Packaging
15664990 6 x 1 L Pack of 6 -
15624990 1 L 1 L Plastic Bottle
15644990 5 L 5 L Plastic Bottle
15654990 5 L 5 L VOLPAC™ CONTAINER
15614990 10 L 10 L VOLPAC™ CONTAINER
15604990 10 L 10 L -
15634990 4 x 5 L Pack of 4 Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Product Code. 15664990 Supplier Honeywell-Fluka Supplier No. 353556X1L
Request Bulk or Custom Formats

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Volumetric, Reag. Ph. Eur., 0.25 M H2SO4 (0.5N)

Solstice's high-quality titration products cover the complete titration workflow and include:

  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use
  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7664-93-9
Molecular Formula H2O4S
Molecular Weight (g/mol) 98.07
MDL Number MFCD00064589
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
Synonym oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
PubChem CID 1118
ChEBI CHEBI:26836
IUPAC Name sulfuric acid
SMILES OS(O)(=O)=O

Specifications

Quantity 6 x 1 L
Linear Formula H2SO4
UN Number UN3264
Formula Weight 98.08
Grade Reagent Ph. Eur.
Chemical Name or Material Sulfuric acid solution, volumetric, 0.25 M H2SO4 (0.5N)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.