missing translation for 'onlineSavingsMsg'
Learn More

Sulfuric Acid Solution 0.05M (0.1N), NIST Standard Concentrate, for Volumetric Analysis, Fisher Chemical™

Product Code. p-7073237 Shop All Fisher Chemical Products
Click to view available options
Quantity:
1 Ampule
6 Ampules
Unit Size:
1 ampoule
6 ampoules
This item is not returnable. View return policy

Product Code. 10325760

Brand: Fisher Chemical J/8460C/05

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

H2O4S, CAS Number-7664-93-9, acide sulfurique, acidum sulfuricum, battery acid, dihydrogen sulfate, dipping acid, electrolyte acid, mattling acid, oil of vitriol, sulphuric acid, vitriol brown oil, 6AMP, 100 deg.C, CHEBI:26836, Colorless, QAOWNCQODCNURD-UHFFFAOYSA-N, sulfuric acid, 64589, 98.072

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7664-93-9
Molecular Formula H2O4S
Molecular Weight (g/mol) 98.07
MDL Number MFCD00064589
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
Synonym oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
PubChem CID 1118
ChEBI CHEBI:26836
IUPAC Name sulfuric acid
SMILES OS(O)(=O)=O

Specifications

pH 1.5
Color Colorless
Boiling Point 100°C
Packaging Ampoule
Vapor Pressure 1mmHg at 20°C
Quantity 1 Ampule
Physical Form Liquid
compliance-icons
Product Identifier
  • Sulfuric acid 0.05M (0.1N) solution (0.5M (1N) Concentrate)
Signal Word
  • Warning
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
Hazard Statement
  • H290-May be corrosive to metals.
Precautionary Statement
  • P234-Keep only in original container.
  • P390-Absorb spillage to prevent material damage.
Supplemental information
  • MIXTURE LIST-Contains : Sulfuric acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.