missing translation for 'onlineSavingsMsg'
Learn More

Sulfuric Acid Min 95%, for Electronic Use (MOS), Fisher Chemical™

Product Code. p-7073587 Shop All Fisher Chemical Products
Click to view available options
Quantity:
2.5 L
Packaging:
HDPE plastic bottle
Plastic Coated Glass bottle
Unit Size:
2.5 L
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Product Code. 10172350

Brand: Fisher Chemical S/9222/PB17

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Sulfuric Acid Min 95%, for Electronic Use (MOS)

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7664-93-9
Molecular Formula H2O4S
Molecular Weight (g/mol) 98.07
MDL Number MFCD00064589
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
Synonym oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
PubChem CID 1118
ChEBI CHEBI:26836
IUPAC Name sulfuric acid
SMILES OS(O)(=O)=O

Specifications

pH 1
Color Colorless
Melting Point 10°C
Boiling Point 290°C
Packaging HDPE plastic bottle
Vapor Pressure 1mmHg 146
Quantity 2.5 L
Viscosity 21mPa at 25°C
Physical Form Liquid
Formula Weight 98.07g/mol
compliance-icons
Product Identifier
  • Sulfuric acid
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.