missing translation for 'onlineSavingsMsg'
Learn More

Sulfuric Acid Min 95%, for Electronic Use (MOS), Fisher Chemical™

Product Code. 12646777 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
2.5 L
Packaging:
HDPE plastic bottle
Plastic Coated Glass bottle
Unit Size:
2.5 L
Product Code. Quantity Packaging unitSize
12646777 2.5 L Plastic Coated Glass bottle 2.5 L
10172350 2.5 L HDPE plastic bottle 2.5 L
2 options
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Product Code. 12646777

Brand: Fisher Chemical S/9222/PC17

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Sulfuric Acid Min 95%, for Electronic Use (MOS)

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7664-93-9
Molecular Formula H2O4S
Molecular Weight (g/mol) 98.07
MDL Number MFCD00064589
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
Synonym oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
PubChem CID 1118
ChEBI CHEBI:26836
IUPAC Name sulfuric acid
SMILES OS(O)(=O)=O

Specifications

pH 1
Color Colorless
Melting Point 10°C
Boiling Point 290°C
Packaging Plastic Coated Glass bottle
Vapor Pressure 1mmHg 146
Quantity 2.5 L
Viscosity 21mPa at 25°C
Physical Form Liquid
Formula Weight 98.07g/mol
compliance-icons
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.