missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfuric acid, Honeywell™
107.07 GBP valid until 2024-05-31
Use promo code "21664" to get your promotional price.
Use promo code "21664" to get your promotional price.
Click to view available options
Quantity:
2.5 L
Packaging:
Plastic Bottle
Description
- We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
Specifications
CAS | 7664-93-9, 7732-18-5 |
Boiling Point | 310°C (1.013 hPa) |
CAS Min % | ≥1.00%, ≥50.00% |
CAS Max % | <5.00%, ≤100.00% |
Packaging | Plastic Bottle |
Quantity | 2.5 L |
Molecular Formula | H2O4S |
Physical Form | Liquid |
MDL Number | MFCD00064589 |
Beilstein | 2037554 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'