missing translation for 'onlineSavingsMsg'
Learn More

Sulfuric Acid, fuming, 20% free SO3 basis, Honeywell™

Product Code. 15655520 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15655520 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15655520 Supplier Honeywell Research Chemicals Supplier No. 435597500G

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Reagent Grade, 20% free SO3 basis

  • We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
    Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
    Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
TRUSTED_SUSTAINABILITY

Specifications

CAS 8014-95-7
Concentration 20% free SO3 basis
Density 1.925g/mL at 25°C
Vapor Pressure 2 mmHg (25°C)
Quantity 500 g
Molecular Formula H2O7S2
Linear Formula H2SO4 · (SO3)x
MDL Number MFCD00137792
UN Number UN1831-class
Beilstein 2037554
Synonym oleum, fuming sulfuric acid, sulfuric acid, fuming, oleum iodisum, sulfuric acid fuming, oleum fuming sulfuric acid, sulfuric acid, mixt with sulfur trioxide, sulfur trioxide, mixt. with sulfuric acid, sulfuric acid mixture with sulfur trioxide
InChI Key HIFJUMGIHIZEPX-UHFFFAOYSA-N
SMILES O=S(=O)=O.OS(O)(=O)=O
IUPAC Name sulfonylideneoxidane; sulfuric acid
Molecular Weight (g/mol) 178.13
PubChem CID 24681
Formula Weight 98.08g/mol
Chemical Name or Material Sulfuric acid, fuming
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.