missing translation for 'onlineSavingsMsg'
Learn More

Sulfuric Acid, DILUT-IT Concentrate, 0.01M, J.T. Baker™

Product Code. 10484131 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
For dilution to 1 L
Packaging:
LDPE ampule
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10484131 For dilution to 1 L LDPE ampule Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Product Code. 10484131 Supplier JT Baker Supplier No. 4704

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Prepare fresh, reliable standard solutions quickly and accurately. Sulfuric Acid, DILUT-IT Concentrate, 0.01M, J.T. Baker™ is a pre-mixed volumetric concentrate for dilution to 1L that is ideal for titrimetry and other forms of wet chemical analysis.

DILUT-IT solutions are relied upon for fast, accurate results in life sciences and pharmaceutical quality control applications. Simply dilute the volumetric concentrate to 1 liter (carefully following instructions provided with the package) and the resulting solution will have an exact normality within ±0.1%.

  • Enhanced productivity: Eliminate expensive, time-consuming steps, such as the selection and analysis of starting reagents, and weighing and standardization of the final solution
  • Space saving: Products require only one-tenth of the space required for ready-made solutions
  • Reduced shipping costs: Spend less on shipping due to the reduced weight and package size of a concentrate

Specifications

Color Colorless
Melting Point 0°C
Boiling Point 100°C
Concentration 0.01mol/L
Density 1g/cm3 at 20°C
Odor Odorless
Packaging LDPE ampule
CAS 7664-93-9
Normality 0.02N
Quantity For dilution to 1 L
Molecular Formula H2SO4
Physical Form Liquid
Synonym Oil of vitriol
Solubility Information Fully miscible in water
Formula Weight 98.08g/mol
Chemical Name or Material Sulfuric acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.