missing translation for 'onlineSavingsMsg'
Learn More

Sulfuric acid, Acculute Standard Volumetric Solution, Final Concentration 2.0N, Thermo Scientific Chemicals

Product Code. 11349299
Change view
Click to view available options
Quantity:
1 Unit
6 Units
Unit Size:
1 unit
6 units
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11349299 6 Units 6 units
11359299 1 Unit 1 unit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Product Code. 11349299 Supplier Thermo Scientific Alfa Aesar Supplier No. 088568.LS
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7664-93-9
Molecular Formula H2O4S
Molecular Weight (g/mol) 98.07
MDL Number MFCD00064589
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
PubChem CID 1118
ChEBI CHEBI:26836
IUPAC Name sulfuric acid
SMILES OS(O)(=O)=O

Specifications

Odor Acrid
Vapor Pressure 23 hPa (17mm Hg) at 20°C
Physical Form Liquid
Color Colorless to Yellow
Quantity 6 Units
Formula Weight 98.08
Concentration or Composition (by Analyte or Components) Sulfuric acid: 55%; Water: 45%
Chemical Name or Material Sulfuric acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.