missing translation for 'onlineSavingsMsg'
Learn More

Sulfuric acid, 99.9999% (metals basis), 92% min, Thermo Scientific Chemicals

Catalog Number 11351608
Click to view available options
Quantity:
10 mL
100 mL
500 mL
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
7664-93-9
H2O4S
98.07
MFCD00064589
QAOWNCQODCNURD-UHFFFAOYSA-N
oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
1118
CHEBI:26836
sulfuric acid
OS(O)(=O)=O
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
7664-93-9
H2O4S
98.07
MFCD00064589
QAOWNCQODCNURD-UHFFFAOYSA-N
oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
1118
CHEBI:26836
sulfuric acid
OS(O)(=O)=O

Sulfuric acid is used as an electrolyte in lead-acid batteries, and a cleaning agent. It is used in mineral processing, fertilizer manufacturing, oil refining, wastewater processing, and chemical synthesis. It is also used in the production of detergents, synthetic resins, dyestuffs, pharmaceuticals, petroleum catalysts, insecticides, antifreeze, processes such as oil well acidizing, aluminum reduction, paper sizing, pigments, enamels, printing inks, coated fabrics and paper. It is also used in the production of explosives, cellophane, acetate and viscose textiles, lubricants, non-ferrous metals and batteries. Sulfuric acid immobilized on silica gel is used as an efficient and benign catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless to Yellow
Concentration ≤92%
Density 1.84 g/mL
Odor Acrid
Quantity 100 mL
Physical Form Liquid
UN Number UN1830
Merck Index 14,8974
Solubility Information Sulfuric acid is miscible with water at all concentrations.
Formula Weight 98.08
Percent Purity 99.9999%
Assay (metals basis)
Chemical Name or Material Sulfuric acid
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'