missing translation for 'onlineSavingsMsg'
Learn More

Sulfuric acid, 72% w/w aq. soln.

Product Code. p-7059911
Click to view available options
Quantity:
1 L
4 L
Unit Size:
1 L
4 L
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Product Code. 15432397

Brand: Thermo Scientific Alfa Aesar 045596.K2

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy
Regler: Denna produkt innehåller en explosiv prekursor enligt förordning (EG) nr 1148/2019. Alla kunder måste fylla i en deklaration före leverans.

CAS: 7664-93-9 | H2O4S | 98.07 g/mol

Sulfuric acid has useful applications in domestic acidic drain cleaners, as a drying agent, as electrolytes in lead-acid batteries and various cleaning agents. It is also useful for mineral processing, fertilizer manufacture, oil refining, waste water processing and chemical synthesis. The sulfuric acid doped poly (m-aminophenol) is used as an aliphatic alcohol vapor sensor material. The catalytic decomposition of sulfuric acid on mixed Cr/Fe oxide shows useful application in sulfur-iodine cycle for hydrogen production.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 7664-93-9
Molecular Formula H2O4S
Molecular Weight (g/mol) 98.07
MDL Number MFCD00064589
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
Synonym oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
PubChem CID 1118
ChEBI CHEBI:26836
IUPAC Name sulfuric acid
SMILES OS(O)(=O)=O

Specifikationer

Color Colorless to Yellow
Concentration 72% w/w aq. soln.
Odor Acrid
Quantity 1 L
Linear Formula H2SO4
Physical Form Liquid
UN Number UN1830
Solubility Information Miscible with water.
Formula Weight 98.08
Chemical Name or Material Sulfuric acid
compliance-icons
Produktbeteckning
  • Sulfuric acid, 72% w/w aqueous solution
Signalord
  • Danger
Riskkategorier
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 A
Faroangivelser
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Skyddsangivelse
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Korrigering av produktinnehåll

Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.

Produkttitel

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.

Tack! Din feedback har skickats. Fisher Scientific arbetar alltid för att förbättra vårt innehåll åt dig. Vi uppskattar din feedback.