missing translation for 'onlineSavingsMsg'
Learn More

Solstice Sulfuric Acid

Product Code. 16645793 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
1000 kg
Unit Size:
1000kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16645793 1000 kg 1000kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16645793 Supplier Solstice Supplier No. 30741S1000KG
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Sulfuric Acid, 95 to 97%.

  • 95 to 97%

Specifications

pH Acidic
Color Colorless
CAS 7664-93-9
Melting Point -14°C to -10°C
Boiling Point 310°C (1,013 hPa)
CAS Min % ≥50.00%
CAS Max % ≤100.00%
Density 1.840 g/cm3 (20°C)
Packaging Returnable Composite IBC
Vapor Pressure Negligible
Flash Point Not applicable
Quantity 1000 kg
Molecular Formula H2O4S
Viscosity No data available
Physical Form Liquid
MDL Number MFCD00064589
UN Number UN1830
Synonym Battery Acid, Hydrogen Sulfate, Oil of Vitriol
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
SMILES OS(O)(=O)=O
IUPAC Name sulfuric acid
Molecular Weight (g/mol) 98.07
Percent Purity 95 to 97%
Grade ACS
Chemical Name or Material Sulfuric acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.