missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfuric Acid, 0.1M, 0.2N, BAKER ANALYZED, J.T. Baker™
Shop All J.T.Baker Products
Click to view available options
Quantity:
10 L
Packaging:
Polycube
Unit Size:
Each
Description
- Standardized potentiometrically against NIST SRM 351, Sodium Carbonate
- Label provides an actual Lot Analysis as a guarantee of purity and consistency
- Analytical methods used for these determinations are based on standard test procedures described in technical publications
- Improvements developed by J.T. Baker are employed to achieve a higher degree of reliability
Specifications
Specifications
| Melting Point | 0°C |
| Boiling Point | 100°C |
| Concentration | 0.1mol/L |
| Density | 1g/cm3 at 20°C |
| CAS | 7664-93-9 |
| Packaging | Polycube |
| Normality | 0.2N |
| Quantity | 10 L |
| Molecular Formula | H2SO4 |
| Viscosity | 1 mPas at 20°C |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction