missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfuric Acid, 0.05M, BAKER ANALYZED, J.T. Baker™
Shop All J.T.Baker Products
Click to view available options
Quantity:
1 L
Packaging:
HDPE
Unit Size:
1 L
Description
- Standardized potentiometrically against NIST SRM 351, Sodium Carbonate
- Label provides an actual Lot Analysis as a guarantee of purity and consistency
- Analytical methods used for these determinations are based on standard test procedures described in technical publications
- Improvements developed by J.T. Baker are employed to achieve a higher degree of reliability
Specifications
Specifications
| CAS | 7664-93-9 |
| Melting Point | 0°C |
| Boiling Point | 100°C |
| Concentration | 0.05mol/L |
| Density | 1g/cm3 at 20°C |
| Heavy Metals (as Pb) | Max. 0.3 ppm |
| Packaging | HDPE |
| Normality | 0.1N |
| Quantity | 1 L |
| Molecular Formula | H2O4S |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction