missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfur Trioxide Triethylamine Complex, TRC
Specifications
Specifications
| Chemical Name or Material | Sulfur Trioxide Triethylamine Complex |
| CAS | 761-01-3 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C6H15NO3S |
| Quantity | 5 g |
| Synonym | Triethylamine compounded with SO3, Triethylamine compounded with sulfur trioxide, N,N-Diethyl-ethanamine compounded with sulfur trioxide, Sulfur Trioxide compounded with triethylamine, NSC 68185, NSC 8168, Sulfur trioxide-triethylamine compound, Sulfur trioxide-triethylamine complex, Triethylamine-sulfur trioxide compound, Triethylamine-sulfur trioxide complex |
| InChI Formula | InChI=1S/C6H15N.O3S/c1-4-7(5-2)6-3;1-4(2)3/h4-6H2,1-3H3; |
| SMILES | CCN(CC)CC.O=S(=O)=O |
| IUPAC Name | N,N-diethylethanamine;sulfur trioxide |
| Molecular Weight (g/mol) | 181.25 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?