missing translation for 'onlineSavingsMsg'
Learn More

Sulfur trioxide pyridine complex, technical, 48-50% active SO3

Product Code. p-3758088 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Unit Size:
100g
2.5kg
25g
500g
This item is not returnable. View return policy

Product Code. 10496950

Brand: Thermo Scientific Acros 132870250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 26412-87-3 | C5H5NO3S | 159.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Lewis acid
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26412-87-3
Molecular Formula C5H5NO3S
Molecular Weight (g/mol) 159.16
MDL Number MFCD00012437
InChI Key UDYFLDICVHJSOY-UHFFFAOYSA-N
Synonym pyridine sulfur trioxide, sulfur trioxide pyridine complex, pyridine; sulfur trioxide, pyridine sulfotrioxide, pyridine compound with sulfur trioxide 1:1, sulfur trioxide, compd. with pyridine, pyridine-sulfur trioxide complex, active sulfure trioxide, sulfur trioxide-pyridine, pyridine-sulfur trioxide
PubChem CID 168533
SMILES O=S(=O)=O.C1=CC=NC=C1

Specifications

Melting Point 155.0°C
Color White to Beige
Infrared Spectrum Authentic
Packaging Plastic bottle
Linear Formula SO3·C5H5N
Quantity 25 g
Beilstein 20,19
Fieser 01,1127; 02,393; 03,275; 04,473; 08,391
Solubility Information Solubility in water: reacts. Other solubilities: soluble in conc. h2so4,hcl and hclo4,soluble in dmf
IUPAC Name pyridine;sulfur trioxide
Formula Weight 159.16
Percent Purity 48-50% active SO3
Grade Technical
Physical Form Powder
Chemical Name or Material Sulfur trioxide pyridine complex, 48.8 to 50.3% active SO3
Show More Show Less
compliance-icons
Product Identifier
  • Sulfur trioxide pyridine complex, 48-50% active SO3
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.