missing translation for 'onlineSavingsMsg'
Learn More

Sulfur trioxide pyridine complex, technical, 48-50% active SO3

Product Code. 10115643 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Unit Size:
100g
2.5kg
25g
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10115643 500 g Plastic bottle 500g
10496950 25 g Plastic bottle 25g
10376730 100 g Plastic bottle 100g
11986291 2.5 kg Plastic bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10115643 Supplier Thermo Scientific Acros Supplier No. 132875000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 26412-87-3 | C5H5NO3S | 159.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Lewis acid
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26412-87-3
Molecular Formula C5H5NO3S
Molecular Weight (g/mol) 159.16
MDL Number MFCD00012437
InChI Key UDYFLDICVHJSOY-UHFFFAOYSA-N
Synonym pyridine sulfur trioxide, sulfur trioxide pyridine complex, pyridine; sulfur trioxide, pyridine sulfotrioxide, pyridine compound with sulfur trioxide 1:1, sulfur trioxide, compd. with pyridine, pyridine-sulfur trioxide complex, active sulfure trioxide, sulfur trioxide-pyridine, pyridine-sulfur trioxide
PubChem CID 168533
SMILES O=S(=O)=O.C1=CC=NC=C1

Specifications

Melting Point 155.0°C
Color White to Beige
Infrared Spectrum Authentic
Packaging Plastic bottle
Linear Formula SO3·C5H5N
Quantity 500 g
Beilstein 20,19
Fieser 01,1127; 02,393; 03,275; 04,473; 08,391
Solubility Information Solubility in water: reacts. Other solubilities: soluble in conc. h2so4,hcl and hclo4,soluble in dmf
IUPAC Name pyridine;sulfur trioxide
Formula Weight 159.16
Percent Purity 48-50% active SO3
Grade Technical
Physical Form Powder
Chemical Name or Material Sulfur trioxide pyridine complex, 48.8 to 50.3% active SO3
Show More Show Less
compliance-icons
Product Identifier
  • Sulfur trioxide pyridine complex, 48-50% active SO3
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.