missing translation for 'onlineSavingsMsg'
Learn More

Sulfur trioxide N,N-dimethylformamide Complex, TRC

Product Code. 30427481
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30427481 1 g
30412011 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30427481 Supplier Toronto Research Chemicals Supplier No. TRCS7894601G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 29584-42-7
Molecular Formula C3H7NO4S
Molecular Weight (g/mol) 153.16
Synonym N,N-Dimethyl-formamide Compd. with sulfur trioxide, N,N-Dimethyl-formamide Compd. with SO3, Sulfur Trioxide Compd. with N,N-dimethylformamide, DMF Compound with Sulfur Trioxide, DMF-sulfur Trioxide Reaction Product, Dimethylformamide Compound with Sulfur Trioxide, Dimethylformamide-sulfur Trioxide Complex, Dimethylformamide-sulfur Trioxide Compound, N,N-Dimethylformamide Compound with Sulfur Trioxide, N,N-Dimethylformamide-sulfur Trioxide Complex
IUPAC Name N,N-dimethylformamide;sulfur trioxide
SMILES CN(C)C=O.O=S(=O)=O

Specifications

Quantity 1 g
InChI Formula InChI=1S/C3H7NO.O3S/c1-4(2)3-5;1-4(2)3/h3H,1-2H3;
Formula Weight 153.0096
Chemical Name or Material Sulfur trioxide N,N-dimethylformamide Complex
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.