missing translation for 'onlineSavingsMsg'
Learn More

Sulfur trioxide N,N-dimethylformamide complex, 47+% active SO3

Product Code. p-3770995 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 10113104

Brand: Thermo Scientific Acros 425850050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 29584-42-7 | C3H7NO4S | 153.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Lewis acid
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 29584-42-7
Molecular Formula C3H7NO4S
Molecular Weight (g/mol) 153.15
MDL Number MFCD00043411
InChI Key AFDQGRURHDVABZ-UHFFFAOYSA-N
Synonym n,n-dimethylformamide sulfur trioxide complex, dmf-sulfur trioxide complex, n,n-dimethylformamide; sulfur trioxide, sulfur trioxide dimethylformamide complex, formamide, n,n-dimethyl-, compd. with sulfur trioxide 1:1, dmf.so3, n,n-dimethylformamide, compound with sulphur trioxide, dimethyl formamide sulfur trioxide, dimethylformamide; sulfur trioxide, sulfur trioxide n,n-dimethylformamide complex
PubChem CID 169056
IUPAC Name N,N-dimethylformamide;sulfur trioxide
SMILES O=S(=O)=O.CN(C)C=O

Specifications

Melting Point 155.0°C to 158.0°C
Color Beige to Brown
Flash Point >120°C
Infrared Spectrum Authentic
Packaging Glass bottle
Linear Formula SO3·HCON(CH3)2
Quantity 5 g
Formula Weight 153.16
Percent Purity 47+% active SO3
Physical Form Crystals
Chemical Name or Material Sulfur trioxide N, N-dimethylformamide complex, 47+% active SO3
Show More Show Less
compliance-icons
Product Identifier
  • Sulfur trioxide N,N-dimethylformamide complex
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.