missing translation for 'onlineSavingsMsg'
Learn More

Sulfur Standard For ICP, 1000μg/mL in Water, SPEX CertiPrep™

Product Code. 16371246 Shop All SPEX Certiprep Products
Change view
Click to view available options
Quantity:
30 mL
125 mL
500 mL
Packaging:
Triple-leached LDPE bottle
Unit Size:
125mL
30mL
500mL
Product Code. Quantity Packaging unitSize
16371246 30 mL Triple-leached LDPE bottle 30mL
11963723 125 mL Triple-leached LDPE bottle 125mL
11953723 500 mL Triple-leached LDPE bottle 500mL
3 options
This item is not returnable. View return policy

Product Code. 16371246

Brand: SPEX Certiprep PLS92M

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
S1000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Chemical Identifiers

CAS 7783-20-2 , 7732-18-5
Molecular Formula H8N2O4S
Molecular Weight (g/mol) 132.13
MDL Number MFCD00003391
InChI Key BFNBIHQBYMNNAN-UHFFFAOYSA-N
PubChem CID 6097028
ChEBI CHEBI:62946
SMILES N.N.OS(O)(=O)=O

Specifications

Packaging Triple-leached LDPE bottle
CAS Min % 1.0, 99.0
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
CAS Max % 1.0, 99.0
Technique AA, ICP
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Type Certified Reference Material
IUPAC Name sulfuric acid diamine
Solvent or Matrix Water
Quantity 30 mL
Grade Assurance
Concentration or Composition (by Analyte or Components) Matrix    Concentration    Cat. No.
H2O    1000μg/mL=10,000=PLS9-2Y, -2XPLS9-3Y, -3X
Chemical Name or Material Sulfur Standard
Show More Show Less

Product intended for laboratory use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.