missing translation for 'onlineSavingsMsg'
Learn More

Sulfur, Reagent grade, Purified by sublimation, Honeywell™

Product Code. 15651980 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 kg
Packaging:
Poly Bottle
Unit Size:
25g
2kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15651980 25 g Poly Bottle 25g
15671980 500 g Poly Bottle 500g
15661980 2 kg Poly Bottle 2kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15651980 Supplier Honeywell Research Chemicals Supplier No. 21523625G
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Reagent Grade, purified by sublimation, -100 mesh particle size, powder

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7704-34-9
Molecular Formula S
Molecular Weight (g/mol) 32.06
MDL Number MFCD00085316
InChI Key NINIDFKCEFEMDL-UHFFFAOYSA-N
Synonym polymeric, lac, ad usum externum, acmc-20a47y, rubbervulcanizationagentis ot
PubChem CID 5362487
ChEBI CHEBI:26833
IUPAC Name sulfur
SMILES [S]

Specifications

Boiling Point 445°C
Melting Point 120°C
Color Yellow
Physical Form Powder
Quantity 25 g
UN Number UN1350
Flash Point 207°C (404.6°F)
Formula Weight 32.07g/mol
Grade Reagent
Ignition Point 450°F
Packaging Poly Bottle
Vapor Pressure 1mmHg (183.8°C), 10mmHg (246°C)
Density 2.07g/cu.cm; Vapor Density: 8.9 (vs air)
Chemical Name or Material Sulfur, Purified by sublimation
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.