missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfur, plasma standard solution, Specpure™ S 10,000μg/mL, Thermo Scientific Chemicals
£58.00 - £201.00
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11312388
|
Thermo Scientific Alfa Aesar
014436.AD |
50 mL |
£58.00
50mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11322388
|
Thermo Scientific Alfa Aesar
014436.AE |
100 mL |
£82.90
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11450460
|
Thermo Scientific Alfa Aesar
014436.AP |
500 mL |
£201.00
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
MFCD00085316 |
Specifications
MFCD00085316 | |
Specpure™ | |
Liquid | |
Sulfur plasma standard solution, S 10000μg/mL |
Safety and Handling
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only