missing translation for 'onlineSavingsMsg'
Learn More

Sulfur pieces, 99.99% (metals basis), Alfa Aesar™, Puratronic™

Brand:  Alfa Aesar 010755.18

Code : NEW

Additional Details : CAS Number : 7704-34-9 Weight : 0.05000kg Transport : UN number : 1350 Chem class : 1 Pack group : III

Quantity 50g

 View more versions of this product

Product Code. 11325809

  • £77.20 / 50g


Estimated Shipment date
from Supplier 23-02-2018
Add to basket

Specifications

Chemical Identifiers

Sulphur
Pieces
MFCD00085316
7704-34-9
99.999% (Metals basis)

Specifications

Sulphur
Pieces
Pale yellow pieces
2.07g/cm3
Pale yellow
7704-34-9
99.999% (Metals basis)
MFCD00085316
Weak
50g
Videos
SDS