missing translation for 'onlineSavingsMsg'
Learn More

Sulfur Oil Standards for Diesel Fuel Analysis, SPEX CertiPrep™

Product Code. 16381186 Shop All SPEX Certiprep Products
Change view
Click to view available options
Concentration:
10 μg/g
100 μg/g
1000 μg/g
200 μg/g
25 μg/g
300 μg/g
5 μg/g
50 μg/g
75 μg/g
750 μg/g
Unit Size:
100g
100mL
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Concentration Unit Size
16381186 300 μg/g 100mL
11955094 5 μg/g 100g
11915094 10 μg/g 100g
11945094 25 μg/g 100g
16371186 50 μg/g 100mL
11965094 75 μg/g 100g
16351186 100 μg/g 100mL
15688285 200 μg/g 100mL
16341186 750 μg/g 100mL
16361186 1000 μg/g 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
This item is not returnable. View return policy
Product Code. 16381186 Supplier SPEX Certiprep Supplier No. DSS8CY
  • ONLINE EXCLUSIVE PRICE
    £109.00
    £96.10 / 100mL
    Save £12.90
    11% Off
 Restricted Item
Estimated Shipment: 24-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
S300μg/g

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Chemical Identifiers

CAS 7704-34-9 , 8042-47-5
Molecular Formula S
Molecular Weight (g/mol) 32.06
MDL Number MFCD00085316
InChI Key NINIDFKCEFEMDL-UHFFFAOYSA-N
PubChem CID 5362487
ChEBI CHEBI:26833
IUPAC Name sulfur
SMILES [S]

Specifications

Packaging Triple-leached LDPE bottle
CAS Min % 0.0, 99.99
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
CAS Max % 0.0, 99.99
Physical Form Solid
Concentration 300 μg/g
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Type Certified Reference Material
Solvent or Matrix Base Oil 20
Industry Type Petrochemical Testing
Quantity 100 mL
Chemical Name or Material Sulfur Oil Standard in Base Oil
Show More Show Less
compliance-icons
Product Identifier
  • Sulfur Oil Standard in Base OIl 20
Signal Word
  • Warning
Hazard Category
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P321-Specific treatment (see on this label).
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Mineral Oil

Product intended for laboratory use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.