missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfur Oil Standards for Diesel Fuel Analysis in Base Oil, Set of 8, SPEX CertiPrep™
Shop All SPEX Certiprep Products
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
Description
Includes
NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
- Sulfur Oil Standard Set
- Includes sulfur standards in Base Oil 20 in concentrations of 50, 100, 200, 300, 500, 750 and 1,000 μg/g
- Also includes Base Oil Blank
- 100 mL volume for each component in the set
Certifications
ISO/IEC 17025:2005, ISO 17034:2016, ISO 9001: 2015
Compliance
GHS/OSHA Complaint
Specifications
Specifications
| Chemical Name or Material | Sulfur Oil Standards in Base Oil |
| Name Note | Set of 7 Standards and Base Oil 20 Blank |
| Packaging | Triple-leached LDPE bottle |
| Certificate of Analysis | With Certificate of Analysis. |
| Concentration or Composition Notes | Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. |
| For Use With (Equipment) | AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation |
| Type | Certified Reference Material |
| Solvent or Matrix | Base Oil 20 |
| Industry Type | Petrochemical Testing |
| Quantity | 1 Kit |
| Show More |
Safety and Handling
Product Identifier
- Sulfur Oil Standards for Diesel Fuel Analysis
- Warning
- Skin sensitiser Category 1
- H317-May cause an allergic skin reaction.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P321-Specific treatment (see on this label).
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : Mineral Oil
Product intended for laboratory use only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction