missing translation for 'onlineSavingsMsg'
Learn More

Sulfur Oil Standard in #2 Diesel Fuel, 20μg/g, SPEX CertiPrep™

Product Code. 15296316 Shop All SPEX Certiprep Products
Click to view available options
Quantity:
100 mL
Unit Size:
100g
This item is not returnable. View return policy

Product Code. 15296316

Brand: SPEX Certiprep SDFS20Y

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
S20μg/g

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Packaging Triple-leached LDPE bottle
CAS 7704-34-9, 68476-34-6
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
CAS Max % 100.0, 0.0
Analyte or Component Names Sulphur
Molecular Formula S
MDL Number MFCD00085316
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
InChI Key NINIDFKCEFEMDL-UHFFFAOYSA-N
Type Certified Reference Material
SMILES [S]
Element Sulphur
IUPAC Name sulfur
Solvent or Matrix #2 Diesel Fuel
Industry Type Petrochemical Testing
Molecular Weight (g/mol) 32.06
PubChem CID 5362487
Quantity 100 mL
ChEBI CHEBI:26833
Concentration or Composition (by Analyte or Components) 20μg/g
Chemical Name or Material Sulfur Oil Standard in #2 Diesel Fuel, For Diesel Fuel Analysis
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Sulfur Oil Standard in 2 Diesel Fuel
Signal Word
  • Warning
Hazard Category
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H351-Suspected of causing cancer.
Precautionary Statement
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Fuels, diesel, No 2

Product intended for laboratory use only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.