missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfur & Metals in Oil Standard 5: S @ 4.5 wt%, Fe @ 250 μg/g, Ni @ 60 μg/g, V @ 100 μg/g in 20 cSt Mineral Oil, VHG™
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
Description
Sulfur & Metals in Oil Standard 5: S @ 4.5 wt%, Fe @ 250 μg/g, Ni @ 60 μg/g, V @ 100 μg/g in 20 cSt Mineral Oil
Specifications
Specifications
| Chemical Name or Material | Sulfur/Nickel/Vanadium/Iron |
| CAS | 7704-34-9, 7440-02-0, 7440-62-2, 7439-89-6 |
| Methods | XRF |
| Analyte or Component Names | Sulfur/Nickel/Vanadium/Iron |
| Shipping Condition | Room Temperature |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | 20 cSt Mineral Oil |
| Quantity | 100 mL |
| Concentration or Composition (by Analyte or Components) | Sulfur 4.5%/Nickel 60 μg/g/Vanadium 100 μg/g/Iron 250 μg/g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction