missing translation for 'onlineSavingsMsg'
Learn More

Sulfur in Kerosene standard solution, Specpure™, 50μg/g (0.005%)

Product Code. 15456385
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15456385 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15456385 Supplier Thermo Scientific Alfa Aesar Supplier No. 041705.AE
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

MDL Number MFCD00804268
UN Number UN1223
Quantity 100 mL
Grade Specpure
Chemical Name or Material Sulfur in Kerosene standard solution, 50μg/g (0.005%)
compliance-icons
Product Identifier
  • Sulphur in Kerosene standard solution, Specpure, (0.005%)
Signal Word
  • Danger
Hazard Category
  • Aspiration toxicity Category 1
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
Precautionary Statement
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.
Supplemental information
  • MIXTURE LIST-Contain: Petroleum distillates, hydrotreated light, sulphur

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.