missing translation for 'onlineSavingsMsg'
Learn More

Sulfur-Free Yttrium Standard: Y @ 1000 μg/g in Hydrocarbon Oil, VHG™

Product Code. 17940976
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17940976 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17940976 Supplier VHG Labs Supplier No. VHGOSFY1000A50G
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

Sulfur-Free Yttrium Standard: Y @ 1000 μg/g in Hydrocarbon Oil
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Yttrium
CAS 7440-65-5
Methods XRF
Analyte or Component Names Yttrium
Shipping Condition Room Temperature
Molecular Formula Y
Synonym Y
InChI Formula InChI=1S/Y
SMILES [Y]
IUPAC Name yttrium
Solvent or Matrix 20 cSt Hydrocarbon Oil
Molecular Weight (g/mol) 88.906
Quantity 50 g
Formula Weight 88.906 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.