missing translation for 'onlineSavingsMsg'
Learn More

Sulfur-Free Titanium Concentrate: Ti @ approx. 7 wt% in Hydrocarbons, VHG™

Product Code. 30002999
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 30002999

Brand: VHG Labs VHGROSFTI7100G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

Sulfur-Free Titanium Concentrate: Ti @ approx. 7 wt% in Hydrocarbons
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Titanium
CAS 7440-32-6
Methods XRF
Analyte or Component Names Titanium
Molecular Formula Ti
Synonym Oremet, Titanium alloy, CP Titanium, Titanium VT1
InChI Formula InChI=1S/Ti
SMILES [Ti]
IUPAC Name titanium
Solvent or Matrix Hydrocarbon Oil
Molecular Weight (g/mol) 47.867
Quantity 100 g
Formula Weight 47.948 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.