missing translation for 'onlineSavingsMsg'
Learn More

Sulfur-Free Strontium Concentrate: Sr @ approx. 9 wt% in Hydrocarbons, VHG™

Product Code. 30003063
Change view
Click to view available options
Quantity:
25 g
100 g
missing translation for 'unitSize'
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30003063 25 g 25g
30003164 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 30003063 missing translation for 'mfr' VHG Labs missing translation for 'supplierNo' VHGROSFSR925G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

Sulfur-Free Strontium Concentrate: Sr @ approx. 9 wt% in Hydrocarbons
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Strontium
CAS 7440-24-6
Methods XRF
Analyte or Component Names Strontium
Molecular Formula Sr
Synonym Sr
InChI Formula InChI=1S/Sr
SMILES [Sr]
IUPAC Name strontium
Solvent or Matrix Hydrocarbon Oil
Molecular Weight (g/mol) 87.62
Quantity 25 g
Formula Weight 87.906 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.