missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfur-Free Standard 1604: Ca @ 5000; Mg, Mo, P, Zn @ 1600 μg/g in 75 cSt Oil
Click to view available options
Quantity:
400 g
Unit Size:
400g
Specifications
Specifications
| Chemical Name or Material | Calcium∣Magnesium∣Molybdenum∣Phosphorus∣Zinc |
| CAS | 7440-70-2, 7439-95-4, 7439-98-7, 7723-14-0, 7440-66-6 |
| Analyte or Component Names | Calcium∣Magnesium∣Molybdenum∣Phosphorus∣Zinc |
| Shipping Condition | Room Temperature |
| Solution Type | Mixture |
| Element | Multi-element |
| Solvent or Matrix | 75 cSt Oil |
| Industry Type | Industrial QC |
| Quantity | 400 g |
| Concentration or Composition (by Analyte or Components) | Magnesium 1600 μg/g∣Molybdenum 1600 μg/g∣Phosphorus 1600 μg/g∣Zinc 1600 μg/g∣Calcium 5000 μg/g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction