missing translation for 'onlineSavingsMsg'
Learn More

Sulfur-Free Copper Concentrate: Cu @ approx. 3 wt% in Hydrocarbons, VHG™

Product Code. 30003021
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30003021 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30003021 Supplier VHG Labs Supplier No. VHGROSFCU325G
 Restricted Item
Estimated Shipment: 05-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

Sulfur-Free Copper Concentrate: Cu @ approx. 3 wt% in Hydrocarbons
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Thallium
CAS 7440-28-0
Analyte or Component Names Thallium
Shipping Condition Room Temperature
Molecular Formula Tl
Synonym Ramor, Thallium, metallic, Thallium, elemental
InChI Formula InChI=1S/Tl
SMILES [Tl]
IUPAC Name thallium
Solvent or Matrix Hydrocarbon Oil
Molecular Weight (g/mol) 204.38
Quantity 25 g
Formula Weight 204.974 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.