missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfur-Free Barium Concentrate: Ba @ approx. 7 wt% in Hydrocarbons, VHG™
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
Description
Sulfur-Free Barium Concentrate: Ba @ approx. 7 wt% in Hydrocarbons
Specifications
Specifications
| Chemical Name or Material | Palladium |
| CAS | 5-3-7440 |
| Methods | XRF |
| Analyte or Component Names | Palladium |
| Molecular Formula | Pd |
| Synonym | Palladium on carbon, Palladium Black, Paladio, Palladium/carbon, Palladium, IG 0218A, MPP 030, MPP 050, MPP 080, P 50, P 50 (metal), Palladex 600, Palladium (nanoparticles), Palladium (powder), Palladium element, SFP 1001P, TCP 211 |
| InChI Formula | InChI=1S/Pd |
| SMILES | [Pd] |
| IUPAC Name | palladium |
| Solvent or Matrix | Hydrocarbon Oil |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction