missing translation for 'onlineSavingsMsg'
Learn More

Sulfur Dioxide Solution, Honeywell Fluka™

Product Code. p-7071938 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
This item is not returnable. View return policy

Product Code. 15652690

Brand: Honeywell-Fluka 247626100G

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 7782-99-2
Molecular Formula H2O3S
Molecular Weight (g/mol) 82.073
MDL Number MFCD00011335
InChI Key LSNNMFCWUKXFEE-UHFFFAOYSA-N
Synonym sulphurous acid, sulfur dioxide solution, schweflige saure german, h2so3, unii-j1p7893f4j, schweflige saure, acide sulfureux, acido sulfuroso, schweflige saeure, trioxosulfuric acid
PubChem CID 1100
ChEBI CHEBI:48854
IUPAC Name sulfurous acid
SMILES OS(=O)O

Specifications

Quantity 100 g
Linear Formula H2SO3
UN Number 1833
Formula Weight 82.08g/mol
Percent Purity ≥6%
Grade ACS Reagent
Residue after Ignition ≤0.005%
Density 1.03g/mL at 25°C; Vapor Density: 2.3 (vs air)
Chemical Name or Material Sulfur Dioxide Solution
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.