missing translation for 'onlineSavingsMsg'
Learn More

Sulfur Certified Reference Material, UKAS ISO 17025 & ISO 17034 Certified, Sulfur in Synthetic Diesel, Blank. 100mL., Paragon Scientific™

Product Code. 16030170
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
This item is not returnable. View return policy

Product Code. 16030170

Brand: Paragon Scientific GOSUBLK

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Paragon Scientific’s Synthetic Diesel Sulfur CRMs are suitable for calibration, verification or for use in quality control procedures used for analysing sulfur content in petroleum products, with sulfur content ranging from 0 (Blank) through to 5.0000% (50000ppm) for each material.

  • Certified in strict accordance to ISO 17025 and ISO 17034 under our UKAS accreditation.
  • Highest level of accreditation guarantee, providing the most credible certified data available worldwide.
  • Suitable for but not limited to internationally recognised test methods and protocol: ASTM D2622, ASTM D4294, ASTM D5453, ASTM D7039, IP 336, IP 496, IP 497.
  • Fully traceable to international standards and low levels of uncertainty achieves maximum accuracy of data.
  • Supplied in 100 mL volume, tamper-evident glass packaging, providing assurance of sample integrity.
  • Manufactured in the United Kingdom.
  • 12 month shelf life.

Specifications

Quantity 100 mL

Nominal values quoted. Certified Values will vary from batch to batch.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.