missing translation for 'onlineSavingsMsg'
Learn More

Sulfotyrosine Antibody (7C5), Janelia Fluor™ 646, Novus Biologicals™

Product Code. 18421403 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18421403 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18421403 Supplier Novus Biologicals Supplier No. NBP259384JF646

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Sulfotyrosine Monoclonal antibody specifically detects Sulfotyrosine in Nonspecies specific samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen Sulfotyrosine
Applications Western Blot, ELISA, Immunofluorescence
Classification Monoclonal
Clone 7C5
Conjugate Janelia Fluor 646
Formulation 50mM Sodium Borate
Gene Alias sTYR;S-Tyrosine;Sulfated Tyrosine;Sulfotyrosine;Sulphated Tyrosine;Sulphotyrosine;Tyrosine sulfation;Tyrosine Sulphation
Host Species Mouse
Immunogen Synthetic Sulfotyrosine conjugated to Keyhole Limpet Hemocyanin (KLH).
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cellular Signaling
Primary or Secondary Primary
Target Species Non-species specific
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.